trans-4-(4-Morpholinyl)cyclohexanol 98%

Lieferant: Apollo Scientific

Synonyme: trans-4-Morpholinocyclohexanol

OR907526-1G
APOSOR907526-1GEA 121 EUR
APOSOR907526-1G
trans-4-(4-Morpholinyl)cyclohexanol 98%
trans-4-(4-Morpholinyl)cyclohexanol
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20259655
CAS-Nummer: 1228947-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR