Methyl-4-hydroxypicolinat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-hydroxypicolinate

OR907529-1G OR907529-250MG
APOSOR907529-1GEA 364 EUR
APOSOR907529-1G APOSOR907529-250MG
Methyl-4-hydroxypicolinat 98%
Methyl-4-hydroxypicolinat
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025998
CAS-Nummer: 473269-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR