4-Brom-2-methoxybenzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Brom-2-methoxybenzoesäurenitril
OR907624-25G
APOSOR907624-25GEA
60.9
EUR
APOSOR907624-25G
4-Brom-2-methoxybenzonitril 97%
4-Brom-2-methoxybenzonitril
Formel:
C₈H₆BrNO Molecular Weight: 212,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738651 CAS-Nummer: 330793-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...