Ethyl-2-(3-bromophenyl)acetate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(3-bromophenyl)acetate

OR907639-25G
APOSOR907639-25GEA 78.3 EUR
APOSOR907639-25G
Ethyl-2-(3-bromophenyl)acetate 97%
Ethyl-2-(3-bromophenyl)acetate
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243.1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953130
CAS-Nummer: 14062-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR