3-Methoxy-2-naphthaldehyd 98%

Lieferant: Apollo Scientific

OR907835-5G
APOSOR907835-5GEA 112 EUR
APOSOR907835-5G
3-Methoxy-2-naphthaldehyd 98%
3-Methoxy-2-naphthaldehyd
Formel: C₁₂H₁₀O₂
Molecular Weight: 186.2066 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13191799
CAS-Nummer: 56679-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR