2-Cyano-3-methoxynaphthalene 97%

Lieferant: Apollo Scientific

OR907837-1G
APOSOR907837-1GEA 632 EUR
APOSOR907837-1G
2-Cyano-3-methoxynaphthalene 97%
2-Cyano-3-methoxynaphthalene
Formel: C₁₂H₉NO
Molecular Weight: 183,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17012444
CAS-Nummer: 92616-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR