Methyl-4-chloro-3-hydroxybenzoate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-chloro-3-hydroxybenzoate

OR907859-1G
APOSOR907859-1GEA 440 EUR
APOSOR907859-1G
Methyl-4-chloro-3-hydroxybenzoate 98%
Methyl-4-chloro-3-hydroxybenzoate
Formel: C₈H₇ClO₃
Molecular Weight: 186.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505968
CAS-Nummer: 166272-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR