Methyl-3-ethoxybenzoat 98%

Lieferant: Apollo Scientific

OR907890-25G
APOSOR907890-25GEA 108 EUR
APOSOR907890-25G
Methyl-3-ethoxybenzoat 98%
Methyl-3-ethoxybenzoat
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00017263
CAS-Nummer: 108593-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR