methyl 4-cyano-3-methylbenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-cyano-3-methylbenzoate

OR907902-5G
APOSOR907902-5GEA 122 EUR
APOSOR907902-5G
methyl 4-cyano-3-methylbenzoate 95%
methyl 4-cyano-3-methylbenzoate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11655969
CAS-Nummer: 25978-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR