3-Formyl-5-methoxybenzoic acid 95%
Lieferant: Apollo Scientific
OR907908-1G
APOSOR907908-1GEA
632
EUR
APOSOR907908-1G
3-Formyl-5-methoxybenzoic acid 95%
3-Formyl-5-methoxybenzoic acid
Formel:
C₉H₈O₄ Molecular Weight: 180,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11042979 CAS-Nummer: 367519-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...