3-Bromofuran-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR907958-1G OR907958-250MG
APOSOR907958-1GEA 166 EUR
APOSOR907958-1G APOSOR907958-250MG
3-Bromofuran-2-carboxylic acid 95%
3-Bromofuran-2-carboxylic acid
Formel: C₅H₃BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00092761
CAS-Nummer: 14903-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR