4-(Ethylcarbamoyl)benzoic acid 98%

Lieferant: Apollo Scientific

OR907989-1G OR907989-5G
APOSOR907989-1GEA 183 EUR
APOSOR907989-1G APOSOR907989-5G
4-(Ethylcarbamoyl)benzoic acid 98%
4-(Ethylcarbamoyl)benzoic acid
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12173485
CAS-Nummer: 167627-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR