2-(Dimethylamino)-2-methylpropanenitrile 97%

Lieferant: Apollo Scientific

OR908028-1G OR908028-5G
APOSOR908028-1GEA 190 EUR
APOSOR908028-1G APOSOR908028-5G
2-(Dimethylamino)-2-methylpropanenitrile 97%
2-(Dimethylamino)-2-methylpropanenitrile
Formel: C₆H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10005348
CAS-Nummer: 2273-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR