2-Chloro-5-hydroxybenzonitrile 98%
Lieferant: Apollo Scientific
OR908030-1G
APOSOR908030-1GEA
136
EUR
APOSOR908030-1G
2-Chloro-5-hydroxybenzonitrile 98%
2-Chloro-5-hydroxybenzonitrile
Formel:
C₇H₄ClNO Molecular Weight: 153,566 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743462 CAS-Nummer: 188774-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...