4,5-Dibromothiophene-2-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

OR908049-1G OR908049-5G
APOSOR908049-1GEA 43.5 EUR
APOSOR908049-1G APOSOR908049-5G
4,5-Dibromothiophene-2-carboxaldehyde
4,5-Dibromothiophene-2-carboxaldehyde
Formel: C₅H₂Br₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00225242
CAS-Nummer: 38071-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR