Methyl 1,3-benzodiazole-4-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1H-benzo[d]imidazole-4-carboxylate
OR908067-1G
APOSOR908067-1GEA
136
EUR
APOSOR908067-1G
Methyl 1,3-benzodiazole-4-carboxylate 98%
Methyl 1,3-benzodiazole-4-carboxylate
Formel:
C₉H₈N₂O₂ Molecular Weight: 176.172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458706 CAS-Nummer: 37619-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...