(2E)-3,4,4-trimethylpent-2-enamide 97%

Lieferant: Apollo Scientific

OR908070-1G
APOSOR908070-1GEA 319 EUR
APOSOR908070-1G
(2E)-3,4,4-trimethylpent-2-enamide 97%
(2E)-3,4,4-trimethylpent-2-enamide
Formel: C₈H₁₅NO
Molecular Weight: 141,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19206375
CAS-Nummer: 860546-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR