2-Bromo-4-hydroxybenzonitrile 98%
Lieferant: Apollo Scientific
OR908076-1G
OR908076-250MG
APOSOR908076-1GEA
256
EUR
APOSOR908076-1G
APOSOR908076-250MG
2-Bromo-4-hydroxybenzonitrile 98%
2-Bromo-4-hydroxybenzonitrile
Formel:
C₇H₄BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD15527339 CAS-Nummer: 82380-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...