4-hydroxy-2-methylbenzonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-2-methylbenzonitrile

OR908077-1G
APOSOR908077-1GEA 109 EUR
APOSOR908077-1G
4-hydroxy-2-methylbenzonitrile 95%
4-hydroxy-2-methylbenzonitrile
Formel: C₈H₇NO
Molecular Weight: 133,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997502
CAS-Nummer: 14143-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR