(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-1-methylpiperidine Dihydrochloride

OR908084-1G
APOSOR908084-1GEA 23.2 EUR
APOSOR908084-1G
(3R)-1-Methylpiperidin-3-amine dihydrochloride 98%
(3R)-1-Methylpiperidin-3-amine dihydrochloride
Formel: C₆H₁₄N₂·2ClH
Molecular Weight: 187.111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20926141
CAS-Nummer: 1157849-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR