3-Cyano-3-phenylpropanoic acid 96%

Lieferant: Apollo Scientific

OR908094-1G
APOSOR908094-1GEA 128 EUR
APOSOR908094-1G
3-Cyano-3-phenylpropanoic acid 96%
3-Cyano-3-phenylpropanoic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03032627
CAS-Nummer: 14387-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR