4-(2-Methoxyphenyl)-1,2,3,6-tetrahydropyridine 95%

Lieferant: Apollo Scientific

OR908113-1G
APOSOR908113-1GEA 509 EUR
APOSOR908113-1G
4-(2-Methoxyphenyl)-1,2,3,6-tetrahydropyridine 95%
4-(2-Methoxyphenyl)-1,2,3,6-tetrahydropyridine
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689320
CAS-Nummer: 154422-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR