3,6-Dihydro-2H-pyran-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR908119-1G
APOSOR908119-1GEA 106 EUR
APOSOR908119-1G
3,6-Dihydro-2H-pyran-4-carbonitrile 95%
3,6-Dihydro-2H-pyran-4-carbonitrile
Formel: C₆H₇NO
Molecular Weight: 109.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20040386
CAS-Nummer: 105772-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR