N-Methyl-2,3-dihydro-1,4-benzodioxine-2-carboxamide 98%

Lieferant: Apollo Scientific

OR908130-1G
APOSOR908130-1GEA 270 EUR
APOSOR908130-1G
N-Methyl-2,3-dihydro-1,4-benzodioxine-2-carboxamide 98%
N-Methyl-2,3-dihydro-1,4-benzodioxine-2-carboxamide
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461072
CAS-Nummer: 21398-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR