Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 2,3-dihydrobenzo[b][1,4]dioxine-5-carboxylate

OR908135-1G
APOSOR908135-1GEA 294 EUR
APOSOR908135-1G
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate 98%
Methyl-2,3-dihydro-1,4-benzodioxine-5-carboxylate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01546256
CAS-Nummer: 214894-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR