2,3-Dihydro-1,4-benzodioxine-5-carbonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydrobenzo[b][1,4]dioxine-5-carbonitrile

OR908136-1G
APOSOR908136-1GEA 102 EUR
APOSOR908136-1G
2,3-Dihydro-1,4-benzodioxine-5-carbonitrile 98%
2,3-Dihydro-1,4-benzodioxine-5-carbonitrile
Formel: C₉H₇NO₂
Molecular Weight: 161.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01116986
CAS-Nummer: 148703-14-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR