4-Hydroxy-3-methylbenzonitrile 98%

Lieferant: Apollo Scientific

OR908148-5G
APOSOR908148-5GEA 185 EUR
APOSOR908148-5G
4-Hydroxy-3-methylbenzonitrile 98%
4-Hydroxy-3-methylbenzonitrile
Formel: C₈H₇NO
Molecular Weight: 133,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848762
CAS-Nummer: 15777-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR