4-Methoxy-3-methylbenzonitrile 98%
Lieferant: Apollo Scientific
OR908149-1G
APOSOR908149-1GEA
98.6
EUR
APOSOR908149-1G
4-Methoxy-3-methylbenzonitrile 98%
4-Methoxy-3-methylbenzonitrile
Formel:
C₉H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12797513 CAS-Nummer: 53078-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...