4-Hydroxy-N-methylbenzamide 98%

Lieferant: Apollo Scientific

OR908160-1G
APOSOR908160-1GEA 311 EUR
APOSOR908160-1G
4-Hydroxy-N-methylbenzamide 98%
4-Hydroxy-N-methylbenzamide
Formel: C₈H₉NO₂
Molecular Weight: 151.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407464
CAS-Nummer: 27642-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR