N-Butyl-3-hydroxybenzamide 98%

Lieferant: Apollo Scientific

OR908169-1G
APOSOR908169-1GEA 290 EUR
APOSOR908169-1G
N-Butyl-3-hydroxybenzamide 98%
N-Butyl-3-hydroxybenzamide
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11130930
CAS-Nummer: 15789-00-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR