Methyl-3-brom-4-hydroxybenzoat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-bromo-4-hydroxybenzoate

OR908176-25G
APOSOR908176-25GEA 46.4 EUR
APOSOR908176-25G
Methyl-3-brom-4-hydroxybenzoat 98%
Methyl-3-brom-4-hydroxybenzoat
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203850
CAS-Nummer: 29415-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR