4,4,6-Trimethyl-2,3-dihydro-1H-quinoline 95%

Lieferant: Apollo Scientific

OR908183-1G
APOSOR908183-1GEA 1020 EUR
APOSOR908183-1G
4,4,6-Trimethyl-2,3-dihydro-1H-quinoline 95%
4,4,6-Trimethyl-2,3-dihydro-1H-quinoline
Formel: C₁₂H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD12913835
CAS-Nummer: 32640-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR