3-tert-Butyl-4-hydroxybenzoic acid 97%

Lieferant: Apollo Scientific

OR908194-1G OR908194-5G
APOSOR908194-1GEA 88.6 EUR
APOSOR908194-1G APOSOR908194-5G
3-tert-Butyl-4-hydroxybenzoic acid 97%
3-tert-Butyl-4-hydroxybenzoic acid
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00016535
CAS-Nummer: 66737-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR