2-Methylthiophene-3-carbonyl chloride 90%
Lieferant: Apollo Scientific
OR908212-1G
APOSOR908212-1GEA
734
EUR
APOSOR908212-1G
2-Methylthiophene-3-carbonyl chloride 90%
2-Methylthiophene-3-carbonyl chloride
Formel:
C₆H₅ClOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD18417771 CAS-Nummer: 123116-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...