Methyl chromane-2-carboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl chroman-2-carboxylate

OR908213-1G
APOSOR908213-1GEA 111 EUR
APOSOR908213-1G
Methyl chromane-2-carboxylate 96%
Methyl chromane-2-carboxylate
Formel: C₁₁H₁₂O₃
Molecular Weight: 192.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527542
CAS-Nummer: 113771-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR