2-Bromo-5-(methoxycarbonyl)benzoic acid ≥98%

Lieferant: Apollo Scientific

OR908215-250MG
APOSOR908215-250MGEA 0 EUR
APOSOR908215-250MG
2-Bromo-5-(methoxycarbonyl)benzoic acid ≥98%
2-Bromo-5-(methoxycarbonyl)benzoic acid
Formel: C₉H₇BrO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22490660
CAS-Nummer: 1621912-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR