2,5-Dimethoxybenzamid 95%

Lieferant: Apollo Scientific

Synonyme: 2,5-Dimethoxybenzoesäureamid

OR908228-1G
APOSOR908228-1GEA 364 EUR
APOSOR908228-1G
2,5-Dimethoxybenzamid 95%
2,5-Dimethoxybenzamid
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 42020-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR