Ethyl-4,4-imethyl-3-oxovalerate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4,4-dimethyl-3-oxopentanoate, Ethyl Pivaloylacetate, 4,4-Dimethyl-3-oxovaleric Acid Ethyl Ester, Pivaloylacetic Acid Ethyl Ester

OR908242-100G
APOSOR908242-100GEA 202 EUR
APOSOR908242-100G
Ethyl-4,4-imethyl-3-oxovalerate 95%
Ethyl-4,4-imethyl-3-oxovalerate
Formel: C₉H₁₆O₃
Molecular Weight: 172.22 g/mol
Siedepunkt: 88…90 °C/9.75mmHg
Dichte: 0.967 g/cm³ (25 °C)
Flammpunkt: 78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042886
CAS-Nummer: 17094-34-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR