(2-methylnaphthalen-1-yl)boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methylnaphthalene-1-boronic Acid, 2-Methyl-1-naphthaleneboronic acid

OR908295-1G OR908295-250MG
APOSOR908295-1GEA 91.5 EUR
APOSOR908295-1G APOSOR908295-250MG
(2-methylnaphthalen-1-yl)boronic acid 95%
(2-methylnaphthalen-1-yl)boronic acid
Formel: C₁₁H₁₁BO₂
Molecular Weight: 186.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03452758
CAS-Nummer: 103989-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR