4-Ethoxy-2-methoxyphenylboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (4-Ethoxy-2-methoxyphenyl)boronic acid

OR908298-1G
APOSOR908298-1GEA 68.3 EUR
APOSOR908298-1G
4-Ethoxy-2-methoxyphenylboronic acid 95%
4-Ethoxy-2-methoxyphenylboronic acid
Formel: C₉H₁₃BO₄
Molecular Weight: 196,008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037217
CAS-Nummer: 1207443-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR