(5-Bromobenzofuran-2-yl)boronic acid ≥98%

Lieferant: Apollo Scientific

OR908307-1G
APOSOR908307-1GEA 120 EUR
APOSOR908307-1G
(5-Bromobenzofuran-2-yl)boronic acid ≥98%
(5-Bromobenzofuran-2-yl)boronic acid
Formel: C₈H₆BBrO₃
Molecular Weight: 240.846 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801726
CAS-Nummer: 331833-99-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR