3-Chloro-5-formylphenylboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-Chloro-5-formylphenyl)boronic acid

OR908333-1G
APOSOR908333-1GEA 232 EUR
APOSOR908333-1G
3-Chloro-5-formylphenylboronic acid 98%
3-Chloro-5-formylphenylboronic acid
Formel: C₇H₆BClO₃
Molecular Weight: 184.385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038799
CAS-Nummer: 1451393-35-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR