(2-Ethynylphenyl)boronic acid 98%

Lieferant: Apollo Scientific

OR908345-5G
APOSOR908345-5GEA 386 EUR
APOSOR908345-5G
(2-Ethynylphenyl)boronic acid 98%
(2-Ethynylphenyl)boronic acid
Formel: C₈H₇BO₂
Molecular Weight: 145.951 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10697431
CAS-Nummer: 905926-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR