(2-Methoxy-6-methylphenyl)boronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxy-6-methylphenylboronic acid
OR908350-1G
APOSOR908350-1GEA
565
EUR
APOSOR908350-1G
(2-Methoxy-6-methylphenyl)boronic acid 98%
(2-Methoxy-6-methylphenyl)boronic acid
Formel:
C₈H₁₁BO₃ Molecular Weight: 165,982 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11043444 CAS-Nummer: 1567218-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...