2-(2-Methoxyethoxy)phenylboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (2-(2-Methoxyethoxy)phenyl)boronic acid

OR908359-1G
APOSOR908359-1GEA 230 EUR
APOSOR908359-1G
2-(2-Methoxyethoxy)phenylboronic acid 95%
2-(2-Methoxyethoxy)phenylboronic acid
Formel: C₉H₁₃BO₄
Molecular Weight: 196,008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11183165
CAS-Nummer: 1122568-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR