2-Allyloxyphenylboronic acid 98%

Lieferant: Apollo Scientific

OR908360-1G
APOSOR908360-1GEA 632 EUR
APOSOR908360-1G
2-Allyloxyphenylboronic acid 98%
2-Allyloxyphenylboronic acid
Formel: C₉H₁₁BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11183169
CAS-Nummer: 151414-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR