2-Cyano-6-methoxyphenylboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-Cyano-6-methoxyphenylboronic Acid

OR908383-1G
APOSOR908383-1GEA 322 EUR
APOSOR908383-1G
2-Cyano-6-methoxyphenylboronic acid 97%
2-Cyano-6-methoxyphenylboronic acid
Formel: C₈H₈BNO₃
Molecular Weight: 176,965 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696677
CAS-Nummer: 1164100-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR