4-(2-Pyridyl)phenylboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-2-yl)phenylboronic acid, (4-(Pyridin-2-yl)phenyl)boronic acid

OR908412-1G OR908412-5G
APOSOR908412-1GEA 74.1 EUR
APOSOR908412-1G APOSOR908412-5G
4-(2-Pyridyl)phenylboronic acid 95%
4-(2-Pyridyl)phenylboronic acid
Formel: C₁₁H₁₀BNO₂
Molecular Weight: 199.014 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17013972
CAS-Nummer: 170230-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR