4-(2-Pyridyl)phenylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
4-(Pyridin-2-yl)phenylboronic acid, (4-(Pyridin-2-yl)phenyl)boronic acid
OR908412-1G
OR908412-5G
APOSOR908412-1GEA
74.1
EUR
APOSOR908412-1G
APOSOR908412-5G
4-(2-Pyridyl)phenylboronic acid 95%
4-(2-Pyridyl)phenylboronic acid
Formel:
C₁₁H₁₀BNO₂ Molecular Weight: 199.014 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17013972 CAS-Nummer: 170230-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...