3-Isobutylphenylboronic acid 96%
Lieferant: Apollo Scientific
OR908451-1G
APOSOR908451-1GEA
632
EUR
APOSOR908451-1G
3-Isobutylphenylboronic acid 96%
3-Isobutylphenylboronic acid
Formel:
C₁₀H₁₅BO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185670 CAS-Nummer: 153624-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...